Assay Details
Gene Symbol: LOC4350917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761630.2 | XP_015617116.1 | 10 - 11 | 4632 | 81 | |
XM_015761631.2 | XP_015617117.1 | 11 - 12 | 4520 | 81 | ||
XM_015761632.2 | XP_015617118.1 | 11 - 12 | 4560 | 81 | ||
XM_015761633.2 | XP_015617119.1 | 11 - 12 | 4544 | 81 | ||
XM_015761634.2 | XP_015617120.1 | 11 - 12 | 4523 | 81 | ||
XM_015761635.2 | XP_015617121.1 | 11 - 12 | 4500 | 81 | ||
GenBank mRNA | AK062893.1 | - | 8 - 9 | 748 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
