Assay Details
| Gene Symbol: LOC4328238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770002.2 | XP_015625488.1 | 7 - 8 | 4044 | 87 | |
| GenBank mRNA | AK241505.1 | - | 1 - 2 | 169 | 87 | |
More Information
Set Membership: |
|

LOC4328238
Genomic Map