Assay Details
Gene Symbol: LOC4346142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792863.2 | XP_015648349.1 | 1 - 2 | 489 | 67 | |
XM_015792864.2 | XP_015648350.1 | 1 - 2 | 489 | 67 | ||
GenBank mRNA | AK061214.1 | - | 1 - 2 | 434 | 67 | |
AK061387.1 | - | 1 - 2 | 434 | 67 | ||
AK067802.1 | - | 1 - 2 | 435 | 67 | ||
AK104251.1 | - | 1 - 2 | 434 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
