Assay Details
| Gene Symbol: LOC4333666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776529.2 | XP_015632015.1 | 2 - 3 | 1966 | 113 | |
| XM_015776530.2 | XP_015632016.1 | 2 - 3 | 1973 | 113 | ||
| GenBank mRNA | AK065363.1 | - | 2 - 3 | 1893 | 113 | |
| AK065952.1 | - | 2 - 3 | 1901 | 113 | ||
| AK101223.1 | - | 1 - 2 | 610 | 113 | ||
| DQ450951.1 | - | 2 - 3 | 1298 | 113 | ||
| HM991170.1 | - | 2 - 3 | 1298 | 113 | ||
More Information
Set Membership: |
|

LOC4333666
Genomic Map