Assay Details
Gene Symbol: LOC4333666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK065363.1 | - | 5 - 6 | 2577 | 67 |
Gene Symbol: LOC107280463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776531.2 | 1 - 2 | 264 | 67 |
Gene Symbol: LOC107280466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776720.2 | XP_015632206.1 | 2 - 3 | 835 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
