Assay Details
| Gene Symbol: LOC4340647 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788244.2 | XP_015643730.1 | 3 - 4 | 1351 | 97 | |
| XM_015788245.2 | XP_015643731.1 | 3 - 4 | 1318 | 97 | ||
| XM_015788246.2 | XP_015643732.1 | 3 - 4 | 1351 | 97 | ||
| XM_015788247.2 | XP_015643733.1 | 3 - 4 | 1335 | 97 | ||
| GenBank mRNA | AK065783.1 | - | 3 - 4 | 1339 | 97 | |
More Information
Set Membership: |
|

LOC4340647
Genomic Map