Assay Details
Gene Symbol: LOC4340647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788246.2 | XP_015643732.1 | 4 - 5 | 1436 | 151 | |
XM_015788247.2 | XP_015643733.1 | 4 - 5 | 1420 | 151 | ||
GenBank mRNA | AK065783.1 | - | 4 - 5 | 1424 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
