Assay Details
Gene Symbol: LOC4340647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788244.2 | XP_015643730.1 | 8 - 9 | 2709 | 100 | |
XM_015788245.2 | XP_015643731.1 | 8 - 9 | 2676 | 100 | ||
XM_015788246.2 | XP_015643732.1 | 6 - 7 | 2484 | 100 | ||
XM_015788247.2 | XP_015643733.1 | 6 - 7 | 2468 | 100 | ||
GenBank mRNA | AK065783.1 | - | 6 - 7 | 2472 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
