Assay Details
| Gene Symbol: LOC4342471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792394.2 | XP_015647880.1 | 8 - 9 | 2633 | 119 | |
| GenBank mRNA | AB583903.1 | - | 8 - 9 | 2383 | 119 | |
| AK111609.1 | - | 9 - 10 | 2540 | 119 | ||
More Information
Set Membership: |
|

LOC4342471
Genomic Map