Assay Details
Gene Symbol: LOC4343813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792286.2 | XP_015647772.1 | 1 | 951 | 105 | |
GenBank mRNA | AK070771.1 | - | 1 | 952 | 105 | |
AK105918.1 | - | 1 | 957 | 105 | ||
AK106214.1 | - | 1 | 957 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
