Assay Details
| Gene Symbol: LOC4328039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767312.2 | XP_015622798.1 | 1 - 2 | 654 | 105 | |
| GenBank mRNA | AK106946.1 | - | 1 - 2 | 598 | 105 | |
More Information
Set Membership: |
|

LOC4328039
Genomic Map