Assay Details
Gene Symbol: LOC4332059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015773099.2 | XP_015628585.1 | 2 - 3 | 349 | 129 | |
GenBank mRNA | AK069728.1 | - | 2 - 3 | 345 | 129 | |
AK070981.1 | - | 2 - 3 | 341 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
