Assay Details
| Gene Symbol: LOC4340110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788577.2 | XP_015644063.1 | 5 - 6 | 3748 | 102 | |
| XM_015788578.2 | XP_015644064.1 | 5 - 6 | 3884 | 102 | ||
| XM_015788579.2 | XP_015644065.1 | 5 - 6 | 3790 | 102 | ||
| XM_015788580.2 | XP_015644066.1 | 5 - 6 | 3807 | 102 | ||
| XM_015788581.2 | XP_015644067.1 | 5 - 6 | 3748 | 102 | ||
| XM_015788582.2 | XP_015644068.1 | 5 - 6 | 3748 | 102 | ||
| GenBank mRNA | AK120452.1 | - | 5 - 6 | 3737 | 102 | |
More Information
Set Membership: |
|

LOC4340110
Genomic Map