Assay Details
| Gene Symbol: LOC4347183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756135.2 | XP_015611621.1 | 3 - 4 | 775 | 101 | |
| GenBank mRNA | AB177378.1 | - | 3 - 4 | 736 | 101 | |
More Information
Set Membership: |
|

LOC4347183
Genomic Map