Assay Details
| Gene Symbol: LOC4329578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771919.2 | XP_015627405.1 | 4 - 5 | 519 | 92 | |
| GenBank mRNA | AK068919.1 | - | 4 - 5 | 496 | 92 | |
| AK104703.1 | - | 4 - 5 | 495 | 92 | ||
| AM039889.1 | - | 4 - 5 | 424 | 92 | ||
More Information
Set Membership: |
|

LOC4329578
Genomic Map