Assay Details
| Gene Symbol: LOC4329911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770238.2 | XP_015625724.1 | 1 - 2 | 151 | 82 | |
| GenBank mRNA | AK105762.1 | - | 1 - 2 | 126 | 82 | |
| AK120769.1 | - | 1 - 2 | 119 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770238.2 transcripts.
Set Membership: |
|

LOC4329911
Genomic Map