Assay Details
| Gene Symbol: LOC4345577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794931.2 | XP_015650417.1 | 4 - 5 | 1903 | 121 | |
| GenBank mRNA | AK070406.1 | - | 4 - 5 | 1909 | 121 | |
More Information
Set Membership: |
|

LOC4345577
Genomic Map