Assay Details
Gene Symbol: LOC4346372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756890.2 | XP_015612376.1 | 1 - 2 | 1290 | 94 | |
GenBank mRNA | AK064377.1 | - | 1 - 2 | 1088 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
