Assay Details
| Gene Symbol: LOC4352684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764836.2 | XP_015620322.1 | 1 - 2 | 1217 | 82 | |
| GenBank mRNA | AK067863.1 | - | 2 - 3 | 591 | 82 | |
More Information
Set Membership: |
|

LOC4352684
Genomic Map