Assay Details
| Gene Symbol: LOC4326797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766481.2 | XP_015621967.1 | 2 - 3 | 611 | 133 | |
| GenBank mRNA | AK103351.1 | - | 2 - 3 | 612 | 133 | |
| AK112012.1 | - | 2 - 3 | 471 | 133 | ||
More Information
Set Membership: |
|

LOC4326797
Genomic Map