Assay Details
Gene Symbol: LOC4352868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288031.1 | - | 1195 | 153 |
Gene Symbol: LOC4327027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759637.2 | XP_015615123.1 | 4 | 1571 | 153 | |
GenBank mRNA | AK107285.1 | - | 1499 | 153 | ||
AK242245.1 | - | 3368 | 153 | |||
EF122457.1 | - | 4 | 1544 | 153 | ||
EF122458.1 | - | 4 | 1544 | 153 | ||
KY038489.1 | - | 4 | 1559 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
