Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105766.4 | XP_001105766.1 | 713 | 100 | ||
| XM_028833954.1 | XP_028689787.1 | 580 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028833954.1 transcripts.
Set Membership: |
|

LAYN
Genomic Map