Assay Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123259.2 | XP_014978745.1 | 865 | 64 | ||
| GenBank mRNA | JU321939.1 | - | 220 | 64 | ||
| JU477870.1 | - | 218 | 64 | |||
| JV563207.1 | - | 221 | 64 | |||
| JV665819.1 | - | 226 | 64 | |||
More Information
Set Membership: |
|

MVB12A
Genomic Map