Assay Details
| Gene Symbol: DYRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261216.1 | NP_001248145.1 | 603 | 74 | ||
| GenBank mRNA | JU471437.1 | - | 603 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DYRK2| Gene Symbol: DYRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261216.1 | NP_001248145.1 | 603 | 74 | ||
| GenBank mRNA | JU471437.1 | - | 603 | 74 | ||
Set Membership: |
|