Assay Details
| Gene Symbol: MTDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266369.1 | NP_001253298.1 | 1603 | 104 | ||
| XM_015145659.2 | XP_015001145.1 | 1723 | 104 | |||
| XM_015145660.2 | XP_015001146.1 | 1534 | 104 | |||
| XM_028852184.1 | XP_028708017.1 | 1657 | 104 | |||
| XM_028852185.1 | XP_028708018.1 | 1723 | 104 | |||
| XM_028852186.1 | XP_028708019.1 | 1567 | 104 | |||
| XM_028852187.1 | XP_028708020.1 | 1659 | 104 | |||
| XM_028852188.1 | XP_028708021.1 | 1468 | 104 | |||
| GenBank mRNA | JU321225.1 | - | 1534 | 104 | ||
| JU321226.1 | - | 1534 | 104 | |||
| JU321229.1 | - | 1534 | 104 | |||
| JU470512.1 | - | 1604 | 104 | |||
| JV044132.1 | - | 1603 | 104 | |||
| JV633395.1 | - | 1606 | 104 | |||
More Information
Set Membership: |
|

MTDH
Genomic Map