Assay Details
| Gene Symbol: TPMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135620.2 | XP_014991106.1 | 694 | 86 | ||
| XM_015135621.2 | XP_014991107.1 | 579 | 86 | |||
| XM_028847815.1 | XP_028703648.1 | 592 | 86 | |||
| GenBank mRNA | JU332165.1 | - | 556 | 86 | ||
| JV722342.1 | - | 574 | 86 | |||
More Information
Set Membership: |
|

TPMT
Genomic Map