Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257464.1 | NP_001244393.1 | 760 | 70 | ||
| XM_028844977.1 | XP_028700810.1 | 768 | 70 | |||
| XM_028844978.1 | XP_028700811.1 | 675 | 70 | |||
| GenBank mRNA | JU320147.1 | - | 760 | 70 | ||
| JU475071.1 | - | 817 | 70 | |||
| JV045231.1 | - | 744 | 70 | |||
| JV638563.1 | - | 807 | 70 | |||
More Information
Set Membership: |
|

KBTBD2
Genomic Map