Assay Details
| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142242.2 | XP_014997728.2 | 772 | 86 | ||
| XM_015142243.2 | XP_014997729.2 | 778 | 86 | |||
| GenBank mRNA | JU326362.1 | - | 441 | 86 | ||
More Information
Set Membership: |
|

GLDN
Genomic Map