Assay Details
| Gene Symbol: UHRF1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136171.2 | XP_014991657.2 | 1275 | 71 | ||
| GenBank mRNA | JU472580.1 | - | 1105 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UHRF1BP1| Gene Symbol: UHRF1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136171.2 | XP_014991657.2 | 1275 | 71 | ||
| GenBank mRNA | JU472580.1 | - | 1105 | 71 | ||
Set Membership: |
|