Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112593.4 | XP_001112593.2 | 649 | 92 | ||
| GenBank mRNA | JU493558.1 | - | 40 | 92 | ||
| JV520548.1 | - | 321 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TEFM
Genomic Map