Assay Details
| Gene Symbol: SPATA19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834180.1 | XP_028690013.1 | 460 | 69 | ||
| XM_028834181.1 | XP_028690014.1 | 460 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPATA19
Genomic Map