Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015153005.2 | XP_015008491.2 | 770 | 55 | ||
| XM_015153006.2 | XP_015008492.2 | 754 | 55 | |||
| XM_028830005.1 | XP_028685838.1 | 736 | 55 | |||
| XM_028830006.1 | XP_028685839.1 | 735 | 55 | |||
More Information
Set Membership: |
|

MORN3
Genomic Map