Assay Details
Gene Symbol: GRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267002.1 | NP_001253931.1 | 1001 | 79 | ||
XM_015138540.2 | XP_014994026.1 | 807 | 79 | |||
XM_015138541.2 | XP_014994027.1 | 715 | 79 | |||
XM_028848086.1 | XP_028703919.1 | 683 | 79 | |||
GenBank mRNA | JU334172.1 | - | 1016 | 79 | ||
JU334173.1 | - | 1016 | 79 | |||
JU475297.1 | - | 1001 | 79 | |||
JV649402.1 | - | 1033 | 79 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
