Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265640.1 | NP_001252569.1 | 329 | 81 | ||
| GenBank mRNA | JV635817.1 | - | 329 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNNI1| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265640.1 | NP_001252569.1 | 329 | 81 | ||
| GenBank mRNA | JV635817.1 | - | 329 | 81 | ||
Set Membership: |
|