Assay Details
| Gene Symbol: ZYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261048.1 | NP_001247977.1 | 3640 | 71 | ||
| GenBank mRNA | JU336318.1 | - | 1611 | 71 | ||
| JU477142.1 | - | 1536 | 71 | |||
| JV048204.1 | - | 3640 | 71 | |||
| JV628257.1 | - | 1651 | 71 | |||
More Information
Set Membership: |
|

ZYX
Genomic Map