Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152139.2 | XP_015007625.1 | 338 | 63 | ||
| GenBank mRNA | JU322330.1 | - | 198 | 63 | ||
| JU473145.1 | - | 205 | 63 | |||
| JV045306.1 | - | 200 | 63 | |||
| JV637001.1 | - | 205 | 63 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TSPAN31
Genomic Map