Assay Details
| Gene Symbol: HIF1AN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147954.2 | XP_015003440.1 | 820 | 65 | ||
| GenBank mRNA | JU476326.1 | - | 731 | 65 | ||
| JU476327.1 | - | 731 | 65 | |||
| JU476328.1 | - | 731 | 65 | |||
| JU476329.1 | - | 731 | 65 | |||
| JU476330.1 | - | 731 | 65 | |||
| JV047591.1 | - | 748 | 65 | |||
More Information
Set Membership: |
|

HIF1AN
Genomic Map