Assay Details
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257689.1 | NP_001244618.1 | 978 | 115 | ||
XM_028838539.1 | XP_028694372.1 | 1116 | 115 | |||
XM_028838540.1 | XP_028694373.1 | 966 | 115 | |||
GenBank mRNA | JU337184.1 | - | 978 | 115 | ||
JV633944.1 | - | 587 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
