Assay Details
| Gene Symbol: ZNF185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842405.1 | XP_028698238.1 | 2117 | 99 | ||
| XM_028842406.1 | XP_028698239.1 | 2114 | 99 | |||
| XM_028842407.1 | XP_028698240.1 | 2027 | 99 | |||
| XM_028842408.1 | XP_028698241.1 | 2024 | 99 | |||
| XM_028842409.1 | XP_028698242.1 | 1940 | 99 | |||
| XM_028842410.1 | XP_028698243.1 | 1937 | 99 | |||
| GenBank mRNA | JU493066.1 | - | 581 | 99 | ||
| JV722944.1 | - | 610 | 99 | |||
More Information
Set Membership: |
|

ZNF185
Genomic Map