Assay Details
| Gene Symbol: DNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260594.1 | NP_001247523.1 | 225 | 147 | ||
| XM_015144039.2 | XP_014999525.1 | 253 | 147 | |||
| XM_028850217.1 | XP_028706050.1 | 242 | 147 | |||
| GenBank mRNA | JU336609.1 | - | 225 | 147 | ||
More Information
Set Membership: |
|

DNAL1
Genomic Map