Assay Details
Gene Symbol: FHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102451.3 | XP_001102451.2 | 765 | 64 | ||
XM_001102904.3 | XP_001102904.1 | 829 | 64 | |||
XM_001102989.3 | XP_001102989.1 | 695 | 64 | |||
XM_001103156.4 | XP_001103156.2 | 748 | 64 | |||
XM_001103233.3 | XP_001103233.1 | 765 | 64 | |||
XM_002806410.3 | XP_002806456.1 | 833 | 64 | |||
XM_015128282.2 | XP_014983768.1 | 685 | 64 | |||
XM_015128283.1 | XP_014983769.1 | 695 | 64 | |||
XM_015128285.2 | XP_014983771.1 | 748 | 64 | |||
XM_028842116.1 | XP_028697949.1 | 914 | 64 | |||
XM_028842117.1 | XP_028697950.1 | 914 | 64 | |||
XM_028842118.1 | XP_028697951.1 | 685 | 64 | |||
GenBank mRNA | JU319639.1 | - | 627 | 64 | ||
JU319640.1 | - | 627 | 64 | |||
JV044233.1 | - | 712 | 64 | |||
JV667868.1 | - | 807 | 64 | |||
JV667869.1 | - | 807 | 64 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
