Assay Details
| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130349.2 | XP_014985835.1 | 620 | 102 | ||
| XM_028844391.1 | XP_028700224.1 | 431 | 102 | |||
| GenBank mRNA | JU334230.1 | - | 502 | 102 | ||
| JV045653.1 | - | 504 | 102 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LMCD1
Genomic Map