Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265644.1 | NP_001252573.1 | 359 | 63 | ||
| GenBank mRNA | JV636440.1 | - | 359 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A21| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265644.1 | NP_001252573.1 | 359 | 63 | ||
| GenBank mRNA | JV636440.1 | - | 359 | 63 | ||
Set Membership: |
|