Assay Details
| Gene Symbol: LYRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084831.4 | XP_001084831.3 | 619 | 114 | ||
| XM_015125760.2 | XP_014981246.1 | 432 | 114 | |||
| XM_015125761.2 | XP_014981247.1 | 485 | 114 | |||
| XM_015125762.2 | XP_014981248.1 | 346 | 114 | |||
| XM_015125763.2 | XP_014981249.1 | 319 | 114 | |||
| XM_028841398.1 | XP_028697231.1 | 788 | 114 | |||
| XM_028841399.1 | XP_028697232.1 | 267 | 114 | |||
| GenBank mRNA | JU321833.1 | - | 336 | 114 | ||
| JU321834.1 | - | 339 | 114 | |||
| JU473051.1 | - | 353 | 114 | |||
| JV043952.1 | - | 340 | 114 | |||
More Information
Set Membership: |
|

LYRM1
Genomic Map