Assay Details
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265771.1 | NP_001252700.1 | 451 | 68 | ||
GenBank mRNA | JU485524.1 | - | 3935 | 68 | ||
JV667385.1 | - | 415 | 68 | |||
JV667386.1 | - | 451 | 68 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
