Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195659.2 | NP_001182588.2 | 378 | 88 | ||
| XM_015151131.2 | XP_015006617.2 | 633 | 88 | |||
| XM_028828891.1 | XP_028684724.1 | 728 | 88 | |||
| GenBank mRNA | DQ869083.1 | - | 125 | 88 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KCNJ8
Genomic Map