Assay Details
Gene Symbol: UBE2Q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002804883.3 | XP_002804929.2 | 1042 | 99 | ||
XM_015142774.2 | XP_014998260.1 | 1042 | 99 | |||
XM_015142776.2 | XP_014998262.1 | 929 | 99 | |||
XM_015142777.2 | XP_014998263.1 | 937 | 99 | |||
XM_028851169.1 | XP_028707002.1 | 1069 | 99 | |||
XM_028851170.1 | XP_028707003.1 | 1042 | 99 | |||
XM_028851171.1 | XP_028707004.1 | 1069 | 99 | |||
XM_028851172.1 | XP_028707005.1 | 1069 | 99 | |||
XM_028851173.1 | XP_028707006.1 | 956 | 99 | |||
XM_028851174.1 | XP_028707007.1 | 929 | 99 | |||
XM_028851175.1 | XP_028707008.1 | 1069 | 99 | |||
XM_028851177.1 | XP_028707010.1 | 880 | 99 | |||
XM_028851178.1 | XP_028707011.1 | 1069 | 99 | |||
XM_028851179.1 | XP_028707012.1 | 1069 | 99 | |||
GenBank mRNA | JU337111.1 | - | 685 | 99 | ||
JU472746.1 | - | 702 | 99 | |||
JV043150.1 | - | 908 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
