Assay Details
| Gene Symbol: GABRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133643.2 | XP_014989129.1 | 592 | 54 | ||
| XM_028841134.1 | XP_028696967.1 | 1573 | 54 | |||
| XM_028841135.1 | XP_028696968.1 | 1689 | 54 | |||
| GenBank mRNA | JV463472.1 | - | 450 | 54 | ||
| JV577331.1 | - | 568 | 54 | |||
| JV667367.1 | - | 564 | 54 | |||
More Information
Set Membership: |
|

GABRD
Genomic Map