Assay Details
| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149406.2 | XP_015004892.2 | 1050 | 60 | ||
| XM_015149407.2 | XP_015004893.2 | 1050 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TGM6| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149406.2 | XP_015004892.2 | 1050 | 60 | ||
| XM_015149407.2 | XP_015004893.2 | 1050 | 60 | |||
Set Membership: |
|