Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844003.1 | XP_028699836.1 | 579 | 69 | ||
| XM_028844004.1 | XP_028699837.1 | 833 | 69 | |||
| XM_028844005.1 | XP_028699838.1 | 582 | 69 | |||
More Information
Set Membership: |
|

ZNF662
Genomic Map